Ethyl-7-nitroindole-2-carboxylate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 7-nitro-1H-indole-2-carboxylate
H66657-03
H66657-06
AAH66657-03EA
90.1
USD
AAH66657-03
AAH66657-06
Ethyl-7-nitroindole-2-carboxylate 95%
Ethyl-7-nitroindole-2-carboxylate
Purity: 95%
CAS No.: 6960-46-9
Molecular Formula: C11H10N2O4
Formula Weight: 234.21
Physical Form: Solid
Beilstein Reference: 224930
MDL No.: MFCD00216475
Formula:
C₁₁H₁₀N₂O₄ MW: 234.211 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00216475 CAS Number: 6960-46-9 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...