6-Chloro-1H-pyrrolo[2,3-b]pyridine 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
6-Chloro-7-azaindole
H66655-03
H66655-06
AAH66655-03EA
74.1
USD
AAH66655-03
AAH66655-06
6-Chloro-1H-pyrrolo[2,3-b]pyridine 95%
6-Chloro-1H-pyrrolo[2,3-b]pyridine
Purity: 95%
CAS No.: 55052-27-2
Molecular Formula: C7H5ClN2
Formula Weight: 152.58
Physical Form: Powder
Appearance: White to pale brown
Melting Point: 170-175°
Beilstein Reference: 907449
UN No.: 2811
MDL No.: MFCD06659666
Formula:
C₇H₅ClN₂ MW: 152.58 g/mol Melting Pt: 170…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659666 CAS Number: 55052-27-2 UN: 2811 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...