4-Chloroindole-3-acetic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

H66637-03 H66637-06
AAH66637-03EA 151 USD
AAH66637-03 AAH66637-06
4-Chloroindole-3-acetic acid 95%
4-Chloroindole-3-acetic acid

Purity: 95% CAS No.: 2519-61-1 Molecular Formula: C10H8ClNO2 Formula Weight: 209.63 Physical Form: Solid Beilstein Reference: 170659 MDL No.: MFCD00216155

Formula: C₁₀H₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD00216155
CAS Number: 2519-61-1

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR