Methyl-2-((diphenylmethylene)amino)acetate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl 2-((diphenylmethylene)amino)acetate, N-(Diphenylmethylene)glycine methyl ester, Methyl N-(diphenylmethylene)glycinate

H66614-14 H66614-22
AAH66614-14EA 67.3 USD
AAH66614-14 AAH66614-22
Methyl-2-((diphenylmethylene)amino)acetate 98%
Methyl-2-((diphenylmethylene)amino)acetate

Purity: 98% CAS No.: 81167-39-7 Molecular Formula: C16H15NO2 Formula Weight: 253.30 Storage Temperature: 2°C to 8°C Physical Form: Solid Appearance: White to pale yellow Melting Point: 42-44° Beilstein Reference: 9695211 MDL No.: MFCD08272233

Formula: C₁₆H₁₅NO₂
MW: 253.30 g/mol
Melting Pt: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD08272233
CAS Number: 81167-39-7

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR