Methyl Indole-2-carboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66609-03
H66609-06
AAH66609-03EA
38.2
USD
AAH66609-03
AAH66609-06
Methyl Indole-2-carboxylate 97%
Methyl Indole-2-carboxylate
Purity: 97%
CAS No.: 1202-04-6
Molecular Formula: C10H9NO2
Formula Weight: 175.19
Physical Form: Solid
Beilstein Reference: 142184
MDL No.: MFCD00460779
Formula:
C₁₀H₉NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00460779 CAS Number: 1202-04-6 EINECS: 214-861-7 |
Specification Test Results
Purity | Min. 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...