Ethyl-6-chloroindole-2-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 6-chloroindole-2-carboxylate

H66483-03 H66483-06
AAH66483-03EA 47.6 USD
AAH66483-03 AAH66483-06
Ethyl-6-chloroindole-2-carboxylate 97%
Ethyl-6-chloroindole-2-carboxylate

Purity: 97% CAS No.: 27034-51-1 Molecular Formula: C11H10ClNO2 Formula Weight: 223.66 Physical Form: Solid Beilstein Reference: 169178 MDL No.: MFCD03084731

Formula: C₁₁H₁₀ClNO₂
MW: 223.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03084731
CAS Number: 27034-51-1

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR