1H-Indole-2-carbonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Indole-2-carbonitrile
H66451-03
H66451-06
AAH66451-03EA
215
USD
AAH66451-03
AAH66451-06
1H-Indole-2-carbonitrile 97%
1H-Indole-2-carbonitrile
Purity: 97%
CAS No.: 36193-65-4
Molecular Formula: C9H6N2
Formula Weight: 142.16
Physical Form: Solid
Beilstein Reference: 116664
UN No.: 3439
MDL No.: MFCD00187566
Formula:
C₉H₆N₂ MW: 142.161 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00187566 CAS Number: 36193-65-4 UN: 3439 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...