5-Nitroindole-2-carboxylic acid 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66441-03
H66441-06
AAH66441-03EA
57.3
USD
AAH66441-03
AAH66441-06
5-Nitroindole-2-carboxylic acid 96%
5-Nitroindole-2-carboxylic acid
Purity: 96%
CAS No.: 16730-20-4
Molecular Formula: C9H6N2O4
Formula Weight: 206.16
Physical Form: Solid
Beilstein Reference: 203744
MDL No.: MFCD00236372
Formula:
C₉H₆N₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00236372 CAS Number: 16730-20-4 |
Specification Test Results
Assay Minimum | 96 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...