1-Benzylindole-3-carboxylic acid 95%
Supplier: Thermo Scientific Chemicals
H66294-03
H66294-06
AAH66294-03EA
75.6
USD
AAH66294-03
AAH66294-06
1-Benzylindole-3-carboxylic acid 95%
1-Benzylindole-3-carboxylic acid
Purity: 95%
CAS No.: 27018-76-4
Molecular Formula: C16H13NO2
Formula Weight: 251.29
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 404957
MDL No.: MFCD00057094
Formula:
C₁₆H₁₃NO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00057094 CAS Number: 27018-76-4 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...