5-Chloro-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Chloro-7-azaindole

H66268-03 H66268-06
AAH66268-03EA 70 USD
AAH66268-03 AAH66268-06
5-Chloro-1H-pyrrolo[2,3-b]pyridine 95%
5-Chloro-1H-pyrrolo[2,3-b]pyridine

Purity: 95% CAS No.: 866546-07-8 Molecular Formula: C7H5ClN2 Formula Weight: 152.58 Physical Form: Powder Appearance: White to pale brown Melting Point: 161-165° Beilstein Reference: 11085807 UN No.: 2811 MDL No.: MFCD06659676

Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Melting Pt: 161…165 °C
Storage Temperature: Ambient
MDL Number: MFCD06659676
CAS Number: 866546-07-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR