7-Methoxy-2,3-dihydro-1H-inden-1-one 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 7-Methoxy-1-indanone, 7-Methoxy-2,3-dihydro-1H-inden-1-one

H66249-03 H66249-MD
AAH66249-03EA 192 USD
AAH66249-03 AAH66249-MD
7-Methoxy-2,3-dihydro-1H-inden-1-one 95%
7-Methoxy-2,3-dihydro-1H-inden-1-one

Purity: 95% CAS No.: 34985-41-6 Molecular Formula: C10H10O2 Formula Weight: 162.19 Physical Form: Solid Melting Point: 99-102° Beilstein Reference: 907449 MDL No.: MFCD06659666

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Melting Pt: 99…102 °C
Storage Temperature: Ambient
MDL Number: MFCD07784339
CAS Number: 34985-41-6

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR