6-Methoxy-1H-indole-2-carboxylic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Methoxyindole-2-carboxylic acid

H66239-03 H66239-06
AAH66239-03EA 63.2 USD
AAH66239-03 AAH66239-06
6-Methoxy-1H-indole-2-carboxylic acid 95%
6-Methoxy-1H-indole-2-carboxylic acid

Purity: 95% CAS No.: 16732-73-3 Molecular Formula: C10H9NO3 Formula Weight: 191.19 Physical Form: Solid Melting Point: 198-203° Beilstein Reference: 156983 MDL No.: MFCD01548823

Formula: C₁₀H₉NO₃
MW: 191.19 g/mol
Melting Pt: 198…203 °C
Storage Temperature: Ambient
MDL Number: MFCD01548823
CAS Number: 16732-73-3

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR