3-Bromo-1-(phenylsulfonyl)indole 97%

Supplier: Thermo Scientific Chemicals
Warning

H66226-03 H66226-06
AAH66226-03EA 32.6 USD
AAH66226-03 AAH66226-06
3-Bromo-1-(phenylsulfonyl)indole 97%
3-Bromo-1-(phenylsulfonyl)indole

Purity: 97% CAS No.: 99655-68-2 Molecular Formula: C14H10BrNO2S Formula Weight: 336.20 Physical Form: Solid Melting Point: 122-126° Beilstein Reference: 3553533 MDL No.: MFCD02681986

Formula: C₁₄H₁₀BrNO₂S
Melting Pt: 122…126 °C
Storage Temperature: Ambient
MDL Number: MFCD02681986
CAS Number: 99655-68-2

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR