3-Bromo-1H-pyrrolo[2,3-b]pyridine 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Bromo-7-azaindole

H66137-03 H66137-06
AAH66137-03EA 80.9 USD
AAH66137-03 AAH66137-06
3-Bromo-1H-pyrrolo[2,3-b]pyridine 96%
3-Bromo-1H-pyrrolo[2,3-b]pyridine

Purity: 96% CAS No.: 74420-15-8 Molecular Formula: C7H5BrN2 Formula Weight: 197.04 Physical Form: Powder Appearance: White to brown Melting Point: 185-189° Beilstein Reference: 119335 UN No.: 2811 MDL No.: MFCD02179276

Formula: C₇H₅BrN₂
MW: 197.04 g/mol
Melting Pt: 185…189 °C
Storage Temperature: Ambient
MDL Number: MFCD02179276
CAS Number: 74420-15-8
UN: 2811
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR