4-Bromo-7-azaindole 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Bromo-7-azaindole, 4-Bromo-1H-pyrrolo[2,3-b]pyridine

H66114-03 H66114-06
AAH66114-03EA 96.9 USD
AAH66114-03 AAH66114-06
4-Bromo-7-azaindole 95%
4-Bromo-7-azaindole

Purity: 95% CAS No.: 348640-06-2 Molecular Formula: C7H5BrN2 Formula Weight: 197.04 Physical Form: Solid Appearance: Yellow to orange Melting Point: 178-183° Beilstein Reference: 9695211 UN No.: 2811 MDL No.: MFCD08272233

Formula: C₇H₅BrN₂
Melting Pt: 178…183 °C
Storage Temperature: Ambient
MDL Number: MFCD08272233
CAS Number: 348640-06-2
UN: 2811
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR