6-Methoxy-1H-indole-3-carbaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66028-03
H66028-06
AAH66028-03EA
129
USD
AAH66028-03
AAH66028-06
6-Methoxy-1H-indole-3-carbaldehyde 97%
6-Methoxy-1H-indole-3-carbaldehyde
Purity: 97%
CAS No.: 70555-46-3
Molecular Formula: C10H9NO2
Formula Weight: 175.19
Physical Form: Solid
Beilstein Reference: 133013
MDL No.: MFCD03848058
Formula:
C₁₀H₉NO₂ MW: 175.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03848058 CAS Number: 70555-46-3 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...