γ-Aminobutyric acid-t-butyl ester hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-Butyl 4-aminobutanoate hydrochloride, H-γ-Abu-OtBu·HCl, tert-Butyl 4-aminobutyrate hydrochloride

H65936-03 H65936-06
AAH65936-03EA 60 USD
AAH65936-03 AAH65936-06
γ-Aminobutyric acid-t-butyl ester hydrochloride 97%
gamma-Aminobutyric acid-t-butyl ester hydrochloride

Purity: 97% CAS No.: 58640-01-0 Molecular Formula: C8H17NO2.HCl Formula Weight: 195.69 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00270592

Formula: C₈H₁₈ClNO₂
MW: 195.687 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00270592
CAS Number: 58640-01-0

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR