1-Benzyl D-Glutamate 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
D-Glutamic Acid 1-Benzyl Ester, H-D-Glu-OBzl
H65903-06
H65903-14
AAH65903-03EA
22.53
USD
AAH65903-03
AAH65903-06
AAH65903-14
1-Benzyl D-Glutamate 95%
1-Benzyl-D-glutamate
Purity: 95%
CAS No.: 79338-14-0
Molecular Formula: C12H15NO4
Formula Weight: 237.26
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Off-white
Beilstein Reference: 3208599
Optical Rotation: -11°(c=1 in 1M HCl)
MDL No.: MFCD00237065
Formula:
C₁₂H₁₅NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00237065 CAS Number: 79338-14-0 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...