Z-Asp(OtBu)-Osu 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester 1-(N-succinimidyl) ester
H65816-03
H65816-06
AAH65816-03EA
89.6
USD
AAH65816-03
AAH65816-06
AAH65816-MD
Z-Asp(OtBu)-Osu 95%
Z-Asp(OtBu)-Osu
Purity: 95%
CAS No.: 3338-32-7
Molecular Formula: C20H24N2O8
Formula Weight: 420.42
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
Melting Point: 148-152°
Beilstein Reference: 1558119
Optical Rotation: -20°(c=1 in dioxane)
MDL No.: MFCD00153328
Formula:
C₂₀H₂₄N₂O₈ MW: 420.418 g/mol Melting Pt: 148…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153328 CAS Number: 3338-32-7 EINECS: 222-080-8 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...