D-Propargylglycine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: H-D-Pra-OH, (R)-2-Aminopent-4-ynoic acid, (R)-alpha-Propargylglycine

H65798-03 H65798-06
AAH65798-03EA 82.9 USD
AAH65798-03 AAH65798-06
D-Propargylglycine 97%
D-Propargylglycine

Purity: 97% CAS No.: 23235-03-2 Molecular Formula: C5H7NO2 Formula Weight: 113.12 Physical Form: Powder Appearance: White to off-white Odor: Odorless Melting Point: 205° dec. Optical Rotation: +23° (c=2 in water) MDL No.: MFCD00237407

Formula: C₅H₇NO₂
Melting Pt: 205 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00237407
CAS Number: 23235-03-2

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR