2,4-Dichloro-6-(trifluoromethyl)pyrimidine 95%

Supplier: Thermo Scientific Chemicals
Warning

H65715-03 H65715-06
AAH65715-03EA 71.8 USD
AAH65715-03 AAH65715-06 AAH65715-MD
2,4-Dichloro-6-(trifluoromethyl)pyrimidine 95%
2,4-Dichloro-6-(trifluoromethyl)pyrimidine

Purity: 95% CAS No.: 16097-64-6 Molecular Formula: C5HCl2F3N2 Formula Weight: 216.97 Physical Form: Liquid Density: 1.589 Refractive Index: 1.4640 Appearance: Pale yellow Beilstein Reference: 911050 MDL No.: MFCD09910264

Formula: C₅HCl₂F₃N₂
MW: 216.97 g/mol
Density: 1.589
Storage Temperature: Ambient
MDL Number: MFCD09910264
CAS Number: 16097-64-6

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR