N-Methyl-DL-alanine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Me-DL-Ala-OH, H-N-Me-DL-Ala-OH, N-DL-Me-Ala-OH

H65287-14 H65287-22
AAH65287-14EA 76.03 USD
AAH65287-14 AAH65287-22 AAH65287-36
N-Methyl-DL-alanine 98%
N-Methyl-DL-alanine

Purity: 98% CAS No.: 600-21-5 Molecular Formula: C4H9NO2 Formula Weight: 103.12 Physical Form: Crystalline powder Appearance: White to off-white MDL No.: MFCD00063136

Formula: C₄H₉NO₂
MW: 103.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063136
CAS Number: 600-21-5

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR