4-Chloropicolinamide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Chloropyridine-2-carboxamide
H65265-06
H65265-22
AAH65265-06EA
30.6
USD
AAH65265-06
AAH65265-14
AAH65265-22
4-Chloropicolinamide 97%
4-Chloropicolinamide
Purity: 97%
CAS No.: 99586-65-9
Molecular Formula: C6H5ClN2O
Formula Weight: 156.57
Physical Form: Solid
Appearance: Off-white
Melting Point: 158-162°
Beilstein Reference: 115850
MDL No.: MFCD01085339
Formula:
C₆H₅ClN₂O MW: 156.57 g/mol Melting Pt: 158…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD01085339 CAS Number: 99586-65-9 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...