Diethyl-2-thienylmethylphosphonate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diethyl (thiophen-2-ylmethyl)phosphonate
H65232-03
H65232-06
AAH65232-03EA
86.9
USD
AAH65232-03
AAH65232-06
AAH65232-MD
Diethyl-2-thienylmethylphosphonate 95%
Diethyl-2-thienylmethylphosphonate
Purity: 95%
CAS No.: 2026-42-8
Molecular Formula: C9H15O3PS
Formula Weight: 234.25
Physical Form: Liquid
Refractive Index: 1.5200
Appearance: Pale yellow
Beilstein Reference: 1683411
MDL No.: MFCD00128457
Formula:
C₉H₁₅O₃PS MW: 234.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00128457 CAS Number: 2026-42-8 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...