4-(Methylamino)benzonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

H65148-06 H65148-14
AAH65148-06EA 167 USD
AAH65148-06 AAH65148-14
4-(Methylamino)benzonitrile 97%
4-(Methylamino)benzonitrile

Purity: 97% CAS No.: 4714-62-9 Molecular Formula: C8H8N2 Formula Weight: 132.17 Physical Form: Solid Melting Point: 87-91° Beilstein Reference: 2802435 MDL No.: MFCD00016381

Formula: C₈H₈N₂
Melting Pt: 87…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00016381
CAS Number: 4714-62-9

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR