D-Histidine methyl ester dihydrochloride 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-Methyl 2-amino-3-(1H-imidazol-4-yl)propanoate dihydrochloride, H-D-His-OMe.2HCl

H65096-03 H65096-06 H65096-14
AAH65096-03EA 30.7 USD
AAH65096-03 AAH65096-06 AAH65096-14
D-Histidine methyl ester dihydrochloride 95%
D-Histidine methyl ester dihydrochloride

Purity: 95% CAS No.: 4467-54-3 Molecular Formula: C7H11N3O2.2HCl Formula Weight: 242.10 Storage Temperature: 2°C to 8°C Physical Form: Solid Sensitivity: Hygroscopic Appearance: Off-white to white Beilstein Reference: 6491805 Optical Rotation: -9°(c=2 in water) MDL No.: MFCD00066137

Formula: C₇H₁₁N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00066137
CAS Number: 4467-54-3

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR