Ethyl-1-benzylpyrrolidine-3-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 1-benzylpyrrolidine-3-carboxylate

H64962-06 H64962-14
AAH64962-03EA 48.86 USD
AAH64962-03 AAH64962-06 AAH64962-14
Ethyl-1-benzylpyrrolidine-3-carboxylate 97%
Ethyl-1-benzylpyrrolidine-3-carboxylate

Purity: 97% CAS No.: 5747-92-2 Molecular Formula: C14H19NO2 Formula Weight: 233.31 Physical Form: Liquid Appearance: Pale yellow Beilstein Reference: 398388 MDL No.: MFCD04039852

Formula: C₁₄H₁₉NO₂
MW: 233.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039852
CAS Number: 5747-92-2

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR