Ethyl-2-aminonicotinate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 2-aminonicotinate
H64957-06
H64957-14
H64957-22
AAH64957-06EA
44.3
USD
AAH64957-06
AAH64957-14
AAH64957-22
Ethyl-2-aminonicotinate 98%
Ethyl 2-aminonicotinate
Purity: 98%
CAS No.: 13362-26-0
Molecular Formula: C8H10N2O2
Formula Weight: 166.18
Physical Form: Solid
Melting Point: 90-96° lit.
Beilstein Reference: 124680
MDL No.: MFCD01569452
Formula:
C₈H₁₀N₂O₂ Melting Pt: 90…96 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD01569452 CAS Number: 13362-26-0 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...