2-(4-Bromophenyl)propan-2-ol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(4-Bromophenyl)-2-propanol

H64919-03 H64919-MD
AAH64919-03EA 185 USD
AAH64919-03 AAH64919-MD
2-(4-Bromophenyl)propan-2-ol 98%
2-(4-Bromophenyl)propan-2-ol

Purity: 98% CAS No.: 2077-19-2 Molecular Formula: C9H11BrO Formula Weight: 215.09 Physical Form: Solid Density: 1.356 Refractive Index: 1.5520 Appearance: White Melting Point: 40-45° Boiling Point: 75-85° /0.1mm Beilstein Reference: 1935066 MDL No.: MFCD11870096

Formula: C₉H₁₁BrO
Boiling Pt: 75…85° /0.1 mm
Melting Pt: 40…45 °C
Density: 1.356
Storage Temperature: Ambient
MDL Number: MFCD11870096
CAS Number: 2077-19-2
EINECS: 816-112-5

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR