3-Fluoro-2-formylphenylboronic acid 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (3-Fluoro-2-formylphenyl)boronic acid, 3-Fluoro-2-formylbenzeneboronic acid

H64909-03 H64909-06 H64909-14
AAH64909-03EA 39.9 USD
AAH64909-03 AAH64909-06 AAH64909-14
3-Fluoro-2-formylphenylboronic acid 95%
3-Fluoro-2-formylphenylboronic acid

Purity: 95% CAS No.: 871126-15-7 Molecular Formula: C7H6BFO3 Formula Weight: 167.93 Storage Temperature: 2°C to 8°C Physical Form: Solid Sensitivity: Light Sensitive Melting Point: 123-127° UN No.: 3261 MDL No.: MFCD10697421

Formula: C₇H₆BFO₃
MW: 167.93 g/mol
Melting Pt: 123…127 °C
Storage Temperature: Ambient
MDL Number: MFCD10697421
CAS Number: 871126-15-7
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR