3-(9H-Carbazol-9-yl)phenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(9-Carbazolyl)benzeneboronic acid, 3-(9H-Carbazol-9-yl)benzeneboronic Acid, (3-(9H-Carbazol-9-yl)phenyl)boronic acid
H64898-03
H64898-06
AAH64898-03EA
51.7
USD
AAH64898-03
AAH64898-06
AAH64898-MD
3-(9H-Carbazol-9-yl)phenylboronic acid 98%
3-(9H-Carbazol-9-yl)phenylboronic acid
Purity: 98%
CAS No.: 864377-33-3
Molecular Formula: C18H14BNO2
Formula Weight: 287.13
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Off-white
Beilstein Reference: 15530380
MDL No.: MFCD18072506
Formula:
C₁₈H₁₄BNO₂ MW: 287.13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18072506 CAS Number: 864377-33-3 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...