4-Nitrophenylethylamine hydrochloride 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(4-Nitrophenyl)ethylamine hydrochloride

H64894-03 H64894-06 H64894-14
AAH64894-03EA 32.4 USD
AAH64894-03 AAH64894-06 AAH64894-14
4-Nitrophenylethylamine hydrochloride 98+%
4-Nitrophenylethylamine hydrochloride

Purity: 98+% CAS No.: 29968-78-3 Molecular Formula: C8H10N2O2.HCl Formula Weight: 202.64 Physical Form: Crystalline powder Appearance: Yellow to yellow-green Melting Point: 200° lit. Beilstein Reference: 3568915 MDL No.: MFCD00012900

Formula: C₈H₁₁ClN₂O₂
MW: 202.64 g/mol
Melting Pt: 200 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00012900
CAS Number: 29968-78-3
EINECS: 249-980-3

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR