4-(N,N-Diphenylamino)benzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Diphenylaminobenzaldehyde
H64887-03
H64887-06
H64887-14
AAH64887-03EA
31
USD
AAH64887-03
AAH64887-06
AAH64887-14
4-(N,N-Diphenylamino)benzaldehyde 98%
4-(N,N-Diphenylamino)benzaldehyde
Purity: 98%
CAS No.: 4181-05-9
Molecular Formula: C19H15NO
Formula Weight: 273.33
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Pale yellow
Odor: Odorless
Melting Point: 129-133° lit.
Beilstein Reference: 2732795
MDL No.: MFCD00145131
Formula:
C₁₉H₁₅NO MW: 273.33 g/mol Melting Pt: 129…133 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00145131 CAS Number: 4181-05-9 |
Specification Test Results
Assay Minimum | 98 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...