2-Bromo-5-hydroxybenzaldehyde 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64884-06
H64884-14
H64884-22
AAH64884-06EA
41.9
USD
AAH64884-06
AAH64884-14
AAH64884-22
2-Bromo-5-hydroxybenzaldehyde 95%
2-Bromo-5-hydroxybenzaldehyde
Purity: 95%
CAS No.: 2973-80-0
Molecular Formula: C7H5BrO2
Formula Weight: 201.02
Physical Form: Solid
Melting Point: 130-135°
Beilstein Reference: 2079921
UN No.: 3077
MDL No.: MFCD03033065
Formula:
C₇H₅BrO₂ Melting Pt: 130…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD03033065 CAS Number: 2973-80-0 UN: 3077 ADR: "9",III |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...