2-Bromo-4'-methylpropiophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64881-03
H64881-06
H64881-14
AAH64881-03EA
36.5
USD
AAH64881-03
AAH64881-06
AAH64881-14
2-Bromo-4'-methylpropiophenone 98%
2-Bromo-4'-methylpropiophenone
Purity: 98%
CAS No.: 1451-82-7
Molecular Formula: C10H11BrO
Formula Weight: 227.10
Physical Form: Solid
Beilstein Reference: 2207628
MDL No.: MFCD11131402
Formula:
C₁₀H₁₁BrO Storage Temperature: Ambient |
MDL Number:
MFCD11131402 CAS Number: 1451-82-7 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...