4-Bromo-2-nitroanisole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromo-1-methoxy-2-nitrobenzene

H64877-06 H64877-14 H64877-22
AAH64877-06EA 31.9 USD
AAH64877-06 AAH64877-14 AAH64877-22
4-Bromo-2-nitroanisole 97%
4-Bromo-2-nitroanisole

Purity: 97% CAS No.: 33696-00-3 Molecular Formula: C7H6BrNO3 Formula Weight: 232.03 Storage Temperature: 2°C to 8°C Physical Form: Solid Melting Point: 85-100° Beilstein Reference: 2259903 MDL No.: MFCD00055529

Formula: C₇H₆BrNO₃
Melting Pt: 85…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00055529
CAS Number: 33696-00-3
EINECS: 251-642-5

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR