N,N-Dimethylpropionamide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64850-14
H64850-22
AAH64850-14EA
95
USD
AAH64850-14
AAH64850-22
AAH64850-36
N,N-Dimethylpropionamide 98+%
N,N-Dimethylpropionamide
Purity: 98+%
CAS No.: 758-96-3
Molecular Formula: C5H11NO
Formula Weight: 101.15
Flash Point: 62°(144°F)
Physical Form: Liquid
Density: 0.920
Refractive Index: 1.4400
Appearance: Colorless
Melting Point: -45° lit.
Boiling Point: 174-175° lit.
Beilstein Reference: 1740719
MDL No.: MFCD00009301
Formula:
C₅H₁₁NO MW: 101.15 g/mol Boiling Pt: 174…175° (lit.) Melting Pt: –45 °C (lit.) Density: 0.920 Flash Pt: 62 °C(144 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009301 CAS Number: 758-96-3 EINECS: 212-064-9 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...