Dimethyl-4-chloropyridine-2,6-dicarboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dimethyl 4-chloropyridine-2,6-dicarboxylate, Dimethyl 4-Chloro-2,6-pyridinedicarboxylate, 4-Chloro-2,6-pyridinedicarboxylic acid dimethyl ester

H64841-03 H64841-06 H64841-MD
AAH64841-03EA 127 USD
AAH64841-03 AAH64841-06 AAH64841-MD
Dimethyl-4-chloropyridine-2,6-dicarboxylate 97%
Dimethyl-4-chloropyridine-2,6-dicarboxylate

Purity: 98% CAS No.: 5371-70-0 Molecular Formula: owderlNO4 Formula Weight: 229.62 Physical Form: Solid Appearance: Pale yellow Melting Point: 139-144° Beilstein Reference: 197075 MDL No.: MFCD02612781

Formula: C₉H₈ClNO₄
MW: 229.616 g/mol
Melting Pt: 139…144 °C
Storage Temperature: Ambient
MDL Number: MFCD02612781
CAS Number: 5371-70-0

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR