5-Nitro-2-thiophenecarboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Nitrothiophene-2-carboxylic acid

H64821-06 H64821-14 H64821-22
AAH64821-06EA 49 USD
AAH64821-06 AAH64821-14 AAH64821-22
5-Nitro-2-thiophenecarboxylic acid 98%
5-Nitro-2-thiophenecarboxylic acid

Purity: 98% CAS No.: 6317-37-9 Molecular Formula: C5H3NO4S Formula Weight: 173.14 Physical Form: Solid Appearance: Pale yellow Melting Point: 154-159° Beilstein Reference: 139665 MDL No.: MFCD00159552

Formula: C₅H₃NO₄S
Melting Pt: 154…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00159552
CAS Number: 6317-37-9
EINECS: 228-654-4

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR