2-Amino-5-fluoro-4-methylpyridine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64815-03
H64815-06
AAH64815-03EA
75
USD
AAH64815-03
AAH64815-06
AAH64815-MD
2-Amino-5-fluoro-4-methylpyridine 95%
2-Amino-5-fluoro-4-methylpyridine
Purity: 95%
CAS No.: 301222-66-2
Molecular Formula: C6H7FN2
Formula Weight: 126.13
Physical Form: Solid
Appearance: Off-white
Beilstein Reference: 9694924
MDL No.: MFCD06659479
Formula:
C₆H₇FN₂ MW: 126.13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06659479 CAS Number: 301222-66-2 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...