(2-Methyl-3-nitrophenyl)acetic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methyl-3-nitrophenylacetic Acid

H64810-06 H64810-14
AAH64810-06EA 26.4 USD
AAH64810-06 AAH64810-14 AAH64810-22
(2-Methyl-3-nitrophenyl)acetic acid 96%
(2-Methyl-3-nitrophenyl)acetic acid

Purity: 96% CAS No.: 23876-15-5 Molecular Formula: C9H9NO4 Formula Weight: 195.17 Storage Temperature: 2°C to 8°C Physical Form: Solid Melting Point: 132-136° Beilstein Reference: 2581973 MDL No.: MFCD07782132

Formula: C₉H₉NO₄
MW: 195.17 g/mol
Melting Pt: 132…136 °C
Storage Temperature: Ambient
MDL Number: MFCD07782132
CAS Number: 23876-15-5

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR