Pyrazine-2,5-dicarboxylic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,5-Pyrazinedicarboxylic acid
H64795-03
H64795-06
H64795-MD
AAH64795-03EA
70.3
USD
AAH64795-03
AAH64795-06
AAH64795-MD
Pyrazine-2,5-dicarboxylic acid 95%
Pyrazine-2,5-dicarboxylic acid
Purity: 95%
CAS No.: 122-05-4
Molecular Formula: C6H4N2O4
Formula Weight: 168.11
Physical Form: Solid
Melting Point: 255-260° lit.
Beilstein Reference: 139985
MDL No.: MFCD01630902
Formula:
C₆H₄N₂O₄ MW: 168.108 g/mol Melting Pt: 255…260 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD01630902 CAS Number: 122-05-4 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...