9-Bromo-10-phenylanthracene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64794-03
H64794-06
AAH64794-03EA
63.8
USD
AAH64794-03
AAH64794-06
AAH64794-MD
9-Bromo-10-phenylanthracene 98%
9-Bromo-10-phenylanthracene
Purity: 98%
CAS No.: 23674-20-6
Molecular Formula: C20H13Br
Formula Weight: 333.23
Physical Form: Powder
Appearance: Pale yellow
Melting Point: 154-158°
Beilstein Reference: 2277064
MDL No.: MFCD00230983
Formula:
C₂₀H₁₃Br Melting Pt: 154…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00230983 CAS Number: 23674-20-6 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...