Methyl-5-bromo-2-chloronicotinate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Purity: 98%
CAS No.: 78686-79-0
Molecular Formula: C7H5BrClNO2
Formula Weight: 250.48
Flash Point:
Formula:
C₇H₅BrClNO₂ Melting Pt: 48…53 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03844848 CAS Number: 78686-79-0 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...