2-Bromo-4-fluoropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

H64785-03 H64785-06
AAH64785-03EA 61.6 USD
AAH64785-03 AAH64785-06
2-Bromo-4-fluoropyridine 97%
2-Bromo-4-fluoropyridine

Purity: 97% CAS No.: 357927-50-5 Molecular Formula: C5H3BrFN Formula Weight: 175.99 Storage Temperature: 2°C to 8°C Flash Point: 76°(169°F) Physical Form: Liquid Density: 1.699 Refractive Index: 1.5370 Beilstein Reference: 14158224 MDL No.: MFCD09260909

Formula: C₅H₃BrFN
MW: 175.99 g/mol
Density: 1.699
Flash Pt: 76 °C(169 °F)
Storage Temperature: Ambient
MDL Number: MFCD09260909
CAS Number: 357927-50-5

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR