1,2-Dichloro-4-fluoro-5-nitrobenzene 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4,5-Dichloro-2-fluoronitrobenzene
H64782-06
H64782-14
H64782-22
AAH64782-06EA
38.5
USD
AAH64782-06
AAH64782-14
AAH64782-22
1,2-Dichloro-4-fluoro-5-nitrobenzene 95%
1,2-Dichloro-4-fluoro-5-nitrobenzene
Purity: 95%
CAS No.: 2339-78-8
Molecular Formula: C6H2Cl2FNO2
Formula Weight: 209.99
Flash Point:
Formula:
C₆H₂Cl₂FNO₂ MW: 209.99 g/mol Boiling Pt: 247 °C Melting Pt: 17 °C Density: 1.596 Flash Pt: >113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00075330 CAS Number: 2339-78-8 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...