2-(Chloromethyl)benzothiazole 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-(Chloromethyl)-1,3-benzothiazole
H64781-03
H64781-06
AAH64781-03EA
114
USD
AAH64781-03
AAH64781-06
AAH64781-MD
2-(Chloromethyl)benzothiazole 95%
2-(Chloromethyl)benzothiazole
Purity: 95%
CAS No.: 37859-43-1
Molecular Formula: C8H6ClNS
Formula Weight: 183.65
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Sensitivity: Moisture Sensitive
Melting Point: 34°
Boiling Point: 88-99°/0.3mm
Beilstein Reference: 120805
MDL No.: MFCD05664944
Formula:
C₈H₆CINS MW: 183.65 g/mol Boiling Pt: 88…99 °C/0.3 mm Melting Pt: 34 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD05664944 CAS Number: 37859-43-1 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...