3-Oxocyclobutanecarboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

H64774-06 H64774-14
AAH64774-06EA 60.6 USD
AAH64774-06 AAH64774-14
3-Oxocyclobutanecarboxylic acid 98%
3-Oxocyclobutanecarboxylic acid

Purity: 98% CAS No.: 23761-23-1 Molecular Formula: C5H6O3 Formula Weight: 114.10 Physical Form: Solid Appearance: Off-white Melting Point: 69-70° Beilstein Reference: 2078401 MDL No.: MFCD00100900

Formula: C5H6O3
MW: 114.10 g/mol
Melting Pt: 69…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00100900
CAS Number: 23761-23-1

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR