3-Fluoro-5-iodopyridine 95%

Supplier: Thermo Scientific Chemicals
Warning

H64770-03 H64770-06
AAH64770-03EA 139 USD
AAH64770-03 AAH64770-06 AAH64770-MD
3-Fluoro-5-iodopyridine 95%
3-Fluoro-5-iodopyridine

Purity: 95% CAS No.: 757950-13-3 Molecular Formula: C5H3FIN Formula Weight: 222.99 Storage Temperature: 2°C to 8°C Physical Form: Solid Sensitivity: Light Sensitive Appearance: Off-white Beilstein Reference: 11107918 MDL No.: MFCD08457780

Formula: C₅H₃FIN
Storage Temperature: Ambient
MDL Number: MFCD08457780
CAS Number: 757950-13-3

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR