((Difluoromethyl)sulfonyl)benzene 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Difluoromethyl phenyl sulphone

H64760-03 H64760-06
AAH64760-03EA 73.2 USD
AAH64760-03 AAH64760-06 AAH64760-MD
((Difluoromethyl)sulfonyl)benzene 95%
((Difluoromethyl)sulfonyl)benzene

Purity: 95% CAS No.: 1535-65-5 Molecular Formula: C7H6F2O2S Formula Weight: 192.18 Physical Form: Liquid Density: 1.324 Refractive Index: 1.5000 Appearance: Pale yellow Melting Point: 24-25° Beilstein Reference: 2259218 MDL No.: MFCD01050170

Formula: C₇H₆F₂O₂S
MW: 192.18 g/mol
MDL Number: MFCD01050170
CAS Number: 1535-65-5

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR